Learn More
Sigma Aldrich Fine Chemicals Biosciences D Luciferin sodium salt ly50MG
Catalog No. 502907294
Encompass
D-Luciferin is mainly produced in bioluminescent organisms. It can simply circulate across the blood testis barrier blood brain barrier and the blood placenta barrier. This small molecule is sensitive to light and oxygen. D-Luciferin is also sensitive to moisture in its powder form.
Catalog No. 50-290-7294 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L688250MG
D-Luciferin is mainly produced in bioluminescent organisms. It can simply circulate across the blood testis barrier blood brain barrier and the blood placenta barrier. This small molecule is sensitive to light and oxygen. D-Luciferin is also sensitive to moisture in its powder form.