Learn More
Sigma Aldrich Fine Chemicals Biosciences D Luciferin synthetic50MG
Catalog No. 502914418
Encompass
D-Luciferin is a small molecule present abundantly in bioluminescent organisms. This molecule is sensitive to oxygen and light.
Catalog No. 50-291-4418 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L950450MG
D-Luciferin is a small molecule present abundantly in bioluminescent organisms. This molecule is sensitive to oxygen and light.