Learn More
Cayman Chemical D-Luciferin (potassium salt), Purity: ≥95%, CAS: 115144-35-9, 100 mg
SDP
Catalog No. 502972424
Encompass_Preferred
A chemiluminescent substrate of firefly luciferase which produces light upon oxidative decarboxylation in the presence of ATP
Catalog No. 50-297-2424 Supplier Cayman Chemical Supplier No. 14681100 MG
A chemiluminescent substrate of firefly luciferase which produces light upon oxidative decarboxylation in the presence of ATP