Learn More
Cayman Chemical D-Luciferin (potassium salt), Purity: ≥95%, CAS: 115144-35-9, 500 mg
SDP
Catalog No. 502971961
Encompass_Preferred
A chemiluminescent substrate of firefly luciferase which produces light upon oxidative decarboxylation in the presence of ATP
Catalog No. 50-297-1961 Supplier Cayman Chemical Supplier No. 14681500 MG
A chemiluminescent substrate of firefly luciferase which produces light upon oxidative decarboxylation in the presence of ATP