Learn More
Cayman Chemical D-Luciferin (sodium salt), Purity: ≥95%, CAS: 103404-75-7, 500 mg
SDP
Catalog No. 502972749
Encompass_Preferred
A chemiluminescent substrate of firefly luciferase which produces light upon oxidative decarboxylation in the presence of ATP
Catalog No. 50-297-2749 Supplier Cayman Chemical Supplier No. 14682500 MG
A chemiluminescent substrate of firefly luciferase which produces light upon oxidative decarboxylation in the presence of ATP