Learn More
Cayman Chemical D-Luciferin (sodium salt), Purity: ≥95%, CAS: 103404-75-7, 50 mg
SDP
Catalog No. 502971311
Encompass_Preferred
A chemiluminescent substrate of firefly luciferase which produces light upon oxidative decarboxylation in the presence of ATP
Catalog No. 50-297-1311 Supplier Cayman Chemical Supplier No. 1468250 MG
A chemiluminescent substrate of firefly luciferase which produces light upon oxidative decarboxylation in the presence of ATP