Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-Lys(Z)-Pro-Arg-pNA diacetate | 108963-70-8 | 100.0% | 773.83 | 5 MG
SDP

Catalog No. 5000426204
Encompass_Preferred

D-Lys(Z)-Pro-Arg-pNA diacetate is a chromogenic peptide substrate of activated protein C (APC).

  • Solid appearance
  • White to light yellow color
  • Emission at 460
  • Excitation at 380
  • Sequence shortening {d-Lys(Z)}-P-R-pNA
  • Solubility in DMSO is 25 mg/mL (32.31 mM)
  • Solubility in H2O is 2 mg/mL (2.58 mM)
  • Powder storage at -80°C for 2 years or -20°C for 1 year
  • In solvent storage at -80°C for 6 months or -20°C for 1 month in sealed storage, away from moisture and light
  • Shipping at room temperature in continental US; may vary elsewhere

Catalog No. 50-004-26204 Supplier Medchemexpress LLC Supplier No. HYP0021A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Lys(Z)-Pro-Arg-pNA diacetate is a chromogenic peptide substrate of activated protein C (APC).

  • Solid appearance
  • White to light yellow color
  • Emission at 460
  • Excitation at 380
  • Sequence shortening {d-Lys(Z)}-P-R-pNA
  • Solubility in DMSO is 25 mg/mL (32.31 mM)
  • Solubility in H2O is 2 mg/mL (2.58 mM)
  • Powder storage at -80°C for 2 years or -20°C for 1 year
  • In solvent storage at -80°C for 6 months or -20°C for 1 month in sealed storage, away from moisture and light
  • Shipping at room temperature in continental US; may vary elsewhere

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.