Learn More
Medchemexpress LLC D-Lys(Z)-Pro-Arg-pNA diacetate | 108963-70-8 | 99.9% | 773.83 | 10 MG
SDP
Catalog No. 5000426196
Encompass_Preferred
D-Lys(Z)-Pro-Arg-pNA diacetate, also known as Spectrozyme PCa or Chromozym Pca diacetate, is a chromogenic peptide substrate specifically designed for activated protein C (APC). This product is intended for research use only and is not for human use.
  • Chromogenic peptide substrate for activated protein C (APC).
  • High purity of 99.98%.
  • Available in various quantities (5 mg, 10 mg, 50 mg).
  • Solid appearance, white to light yellow in color.
  • Optimal excitation at 380 nm and emission at 460 nm.
Catalog No. 50-004-26196 Supplier Medchemexpress LLC Supplier No. HYP0021A10MG
D-Lys(Z)-Pro-Arg-pNA diacetate, also known as Spectrozyme PCa or Chromozym Pca diacetate, is a chromogenic peptide substrate specifically designed for activated protein C (APC). This product is intended for research use only and is not for human use.
  • Chromogenic peptide substrate for activated protein C (APC).
  • High purity of 99.98%.
  • Available in various quantities (5 mg, 10 mg, 50 mg).
  • Solid appearance, white to light yellow in color.
  • Optimal excitation at 380 nm and emission at 460 nm.