Learn More
Cayman Chemical D-Lys3-GHRP-6trIfluoroace 10mg
SDP
Catalog No. 502909764
Encompass_Preferred
An antagonist of GHS-R1a (Ki = 4.25 nM); inhibits ghrelin-induced cAMP accumulation in human aortic smooth muscle cells at 20 nM; decreases food intake in Zucker FA/FA lean and fa/fa obese rats at 12 mg/kg
Catalog No. 50-290-9764 Supplier Cayman Chemical Supplier No. 3686210 MG
An antagonist of GHS-R1a (Ki = 4.25 nM); inhibits ghrelin-induced cAMP accumulation in human aortic smooth muscle cells at 20 nM; decreases food intake in Zucker FA/FA lean and fa/fa obese rats at 12 mg/kg