Learn More
Cayman Chemical D-Lysine lactam, Purity: ≥95%, CAS: 28957-33-7, 100 mg
SDP
Catalog No. 502990926
Encompass_Preferred
A chiral building block; has been used in the synthesis of a chiral synthetic antibiotic intermediate, as well as in the stereoselective synthesis of NK receptor antagonists
Catalog No. 50-299-0926 Supplier Cayman Chemical Supplier No. 31684100 MG
A chiral building block; has been used in the synthesis of a chiral synthetic antibiotic intermediate, as well as in the stereoselective synthesis of NK receptor antagonists