Learn More
Medchemexpress LLC D-lysine, N2,N6-bis[(1,1-dimethylethoxy)carbonyl] | 65360-27-2 | MFCD00076960 | ≥95.0% | 346.42 g·mol⁻¹ | C16H30N2O6 | 10 G
SDP
Catalog No. 5000301348
Encompass_Preferred
N2,N6-Bis(tert-butoxycarbonyl)-D-lysine is a Boc-protected derivative of D-lysine used as a reagent and intermediate in peptide and organic synthesis. The dual Boc protection of the alpha and side-chain amine groups allows selective deprotection and stepwise coupling in multi-step syntheses. Supplied for research use with typical purity ≥95%, it is intended for workflows requiring temporary protection of lysine amines.
  • Protects both alpha and side-chain amines during synthesis.
  • Enables selective deprotection strategies in peptide assembly.
  • Suitable for solid-phase and solution-phase peptide synthesis.
  • Supplied with typical purity of ≥95% for research use.
  • Available in multiple pack sizes for lab-scale work.
Catalog No. 50-003-01348 Supplier Medchemexpress LLC Supplier No. HYW01852810G
N2,N6-Bis(tert-butoxycarbonyl)-D-lysine is a Boc-protected derivative of D-lysine used as a reagent and intermediate in peptide and organic synthesis. The dual Boc protection of the alpha and side-chain amine groups allows selective deprotection and stepwise coupling in multi-step syntheses. Supplied for research use with typical purity ≥95%, it is intended for workflows requiring temporary protection of lysine amines.
  • Protects both alpha and side-chain amines during synthesis.
  • Enables selective deprotection strategies in peptide assembly.
  • Suitable for solid-phase and solution-phase peptide synthesis.
  • Supplied with typical purity of ≥95% for research use.
  • Available in multiple pack sizes for lab-scale work.