Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Lysine, Thermo Scientific™

Catalog No. AC469950050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC469950050 5 g
AC469950010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC469950050 Supplier Thermo Scientific Chemicals Supplier No. 469950050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 218°C
Chemical Name or Material D-Lysine
CAS 923-27-3
Boiling Point 311°C
Color Beige to White
Infrared Spectrum Conforms
Packaging Glass Bottle
Molecular Formula C6 H14 N2 O2
MDL Number 00008234
Specific Rotation -12° to -16°
Synonym (R)-2,6-Diaminohexanoic acid
Water ≤5 %
Alpha Vector LYSINE
Specific Rotation Condition (20°C, 589 nm) (c=1 aq. soln.)
Molecular Weight (g/mol) 146.19
Quantity 5 g
Physical Form Crystalline powder
Product Line Life science
Shipping Condition No special requirements
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.