Learn More
Medchemexpress LLC D-Lyxose | 1114-34-7 | 98.0% | 100 MG
SDP
Catalog No. 5000339412
Encompass_Preferred
D-Lyxose is an endogenous metabolite and a rare pentose sugar with significant potential for pharmaceutical synthesis. It serves as a starting material for antitumor agents and acts as a precursor for L-nucleoside analogs used in the development of antiviral drugs. It can also be used as a synthetic intermediate for other rare sugars.
  • Used as a starting material for anti-tumor drugs, such as alpha galactose ceramide immunostimulants.
  • Precursor for L-nucleoside analogs for the development of antiviral drugs.
  • Synthetic intermediate for other rare sugars, such as L-ribose.
  • Intended for research and analytical applications.
Catalog No. 50-003-39412 Supplier Medchemexpress LLC Supplier No. HY128753100MG
D-Lyxose is an endogenous metabolite and a rare pentose sugar with significant potential for pharmaceutical synthesis. It serves as a starting material for antitumor agents and acts as a precursor for L-nucleoside analogs used in the development of antiviral drugs. It can also be used as a synthetic intermediate for other rare sugars.
  • Used as a starting material for anti-tumor drugs, such as alpha galactose ceramide immunostimulants.
  • Precursor for L-nucleoside analogs for the development of antiviral drugs.
  • Synthetic intermediate for other rare sugars, such as L-ribose.
  • Intended for research and analytical applications.