Learn More
Medchemexpress LLC D-Lyxose | 1114-34-7 | 98.0% | 500 MG
SDP
Catalog No. 5000339313
Encompass_Preferred
D-Lyxose is an endogenous metabolite and a rare pentose with significant potential for pharmaceutical synthesis. It serves as a starting material for antitumor agents, a precursor for L-nucleoside analogs in antiviral drug development, and a synthetic intermediate for other rare sugars.
  • Serves as a starting material for antitumor agents (e.g., α-galactosylceramide-based immunostimulants).
  • Serves as a precursor of L-nucleoside analogs for the development of antiviral drugs.
  • Serves as a synthetic intermediate for other rare sugars (e.g., L-ribose).
Catalog No. 50-003-39313 Supplier Medchemexpress LLC Supplier No. HY128753500MG
D-Lyxose is an endogenous metabolite and a rare pentose with significant potential for pharmaceutical synthesis. It serves as a starting material for antitumor agents, a precursor for L-nucleoside analogs in antiviral drug development, and a synthetic intermediate for other rare sugars.
  • Serves as a starting material for antitumor agents (e.g., α-galactosylceramide-based immunostimulants).
  • Serves as a precursor of L-nucleoside analogs for the development of antiviral drugs.
  • Serves as a synthetic intermediate for other rare sugars (e.g., L-ribose).