Learn More
Medchemexpress LLC D-(+)-Malic acid | 636-61-3 | 134.09 | 25 G
SDP
Catalog No. 5000402881
Encompass_Preferred
D-(+)-Malic acid, also known as D-Malic acid, is an active enantiomer of Malic acid. It functions as a competitive inhibitor of L-(-)-malic acid transport. It is utilized in research and analytical applications.
  • Active enantiomer of malic acid.
  • Functions as a competitive inhibitor of L-(-)-malic acid transport.
  • Suitable for research and analytical applications.
Catalog No. 50-004-02881 Supplier Medchemexpress LLC Supplier No. 25GHY20558
D-(+)-Malic acid, also known as D-Malic acid, is an active enantiomer of Malic acid. It functions as a competitive inhibitor of L-(-)-malic acid transport. It is utilized in research and analytical applications.
  • Active enantiomer of malic acid.
  • Functions as a competitive inhibitor of L-(-)-malic acid transport.
  • Suitable for research and analytical applications.