Learn More
Medchemexpress LLC D-(+)-Malic acid | 636-61-3 | 134.09 | 250 MG
SDP
Catalog No. 5000402906
Encompass_Preferred
D-(+)-Malic acid, also known as D-Malic acid, is an active enantiomer of Malic acid. It serves as a competitive inhibitor of L-(-)malic acid transport. This product is an analytical standard of D-(+)-Malic acid, specifically intended for research and analytical applications.
  • Competitive inhibitor of L-(-)malic acid transport
  • Suitable for research and analytical applications
  • Enantiomer of Malic acid
Catalog No. 50-004-02906 Supplier Medchemexpress LLC Supplier No. 250MGHY20558R
D-(+)-Malic acid, also known as D-Malic acid, is an active enantiomer of Malic acid. It serves as a competitive inhibitor of L-(-)malic acid transport. This product is an analytical standard of D-(+)-Malic acid, specifically intended for research and analytical applications.
  • Competitive inhibitor of L-(-)malic acid transport
  • Suitable for research and analytical applications
  • Enantiomer of Malic acid