Learn More
Medchemexpress LLC D-(+)-Malic acid | 636-61-3 | 99.75% | 134.09 | 5 G
SDP
Catalog No. 5000400765
Encompass_Preferred
D-(+)-Malic acid is an active enantiomer of Malic acid and a competitive inhibitor of L-(-)malic acid transport. It is a human endogenous metabolite. It is a solid with a white to off-white appearance.
  • Used as a competitive inhibitor of L-(-)malic acid transport
  • Can be produced by certain bacteria from maleic acid
  • Suitable for various biological activities and research applications
Catalog No. 50-004-00765 Supplier Medchemexpress LLC Supplier No. HY205585G
D-(+)-Malic acid is an active enantiomer of Malic acid and a competitive inhibitor of L-(-)malic acid transport. It is a human endogenous metabolite. It is a solid with a white to off-white appearance.
  • Used as a competitive inhibitor of L-(-)malic acid transport
  • Can be produced by certain bacteria from maleic acid
  • Suitable for various biological activities and research applications