Learn More
Medchemexpress LLC D-(+)-Malic acid | 636-61-3 | 99.8% | 134.09 | 1 ML
SDP
Catalog No. 5000400988
Encompass_Preferred
D-(+)-Malic acid (D-Malic acid) is an active enantiomer of Malic acid that functions as a competitive inhibitor of L(-)malic acid transport. It is an endogenous metabolite.
  • Active enantiomer of malic acid
  • Competitive inhibitor of L(-)malic acid transport
  • Can be produced from maleic acid by certain bacteria
Catalog No. 50-004-00988 Supplier Medchemexpress LLC Supplier No. HY2055810MM/1ML
D-(+)-Malic acid (D-Malic acid) is an active enantiomer of Malic acid that functions as a competitive inhibitor of L(-)malic acid transport. It is an endogenous metabolite.
  • Active enantiomer of malic acid
  • Competitive inhibitor of L(-)malic acid transport
  • Can be produced from maleic acid by certain bacteria