Learn More
Medchemexpress LLC D-Malic acid | 636-61-3 | MFCD00004245 | 134.09 g/mol | 100 MG
SDP
Catalog No. 5000401017
Encompass_Preferred
D-Malic acid, also known as (R)-malic acid, is an optically active form of malic acid with an (R)-configuration. It acts as a conjugate acid of (R)-malate(2-) and is the enantiomer of (S)-malic acid. It has been identified as a metabolite in organisms such as Escherichia coli (strain K12, MG1655), Vaccinium macrocarpon, and Arabidopsis thaliana.
  • Optically active form of malic acid with (R)-configuration
  • Conjugate acid of (R)-malate(2-)
  • Enantiomer of (S)-malic acid
  • Primary hazards include corrosive and irritant properties
  • Utilized as an intermediate, chelating agent, buffering agent, and food additive
  • Identified in Escherichia coli, Vaccinium macrocarpon, and Arabidopsis thaliana
Catalog No. 50-004-01017 Supplier Medchemexpress LLC Supplier No. HY20558R100MG
D-Malic acid, also known as (R)-malic acid, is an optically active form of malic acid with an (R)-configuration. It acts as a conjugate acid of (R)-malate(2-) and is the enantiomer of (S)-malic acid. It has been identified as a metabolite in organisms such as Escherichia coli (strain K12, MG1655), Vaccinium macrocarpon, and Arabidopsis thaliana.
  • Optically active form of malic acid with (R)-configuration
  • Conjugate acid of (R)-malate(2-)
  • Enantiomer of (S)-malic acid
  • Primary hazards include corrosive and irritant properties
  • Utilized as an intermediate, chelating agent, buffering agent, and food additive
  • Identified in Escherichia coli, Vaccinium macrocarpon, and Arabidopsis thaliana