Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Malic acid, MilliporeSigma™ Supelco™

Catalog No. 111015002 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5002 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5002 Supplier MilliporeSigma Supelco Supplier No. 46940U
Add to Cart
Add to Cart
Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 636-61-3
Physical Form Neat
Linear Formula HO2 CCH2 CH(OH)CO2 H
MDL Number MFCD00004245
Melting Point 98°C to 102°C
Synonym D-(+)-Malic acid; (R)-(+)-2-Hydroxysuccinic acid; D-Hydroxybutanedioic acid
Type Food Additive Standard
Quantity 100 mg
Formula Weight 134.09
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.