Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger TCI America D-(+)-Maltose Monohydrate, HPLC Grade, 25 g

Catalog No. 502781437
Encompass_Preferred

D-(+)-Maltose Monohydrate is a disaccharide sugar composed of two glucose units linked by an α(1→4) glycosidic bond. This white, crystalline powder is suitable for use in research, brewing, and fermentation processes as a carbohydrate source.

  • Formula: C12H22O11·H2O
  • Molecular weight: 360.32
  • Purity: ≥98.0% (HPLC)
  • Physical state: White powder to crystal
  • Melting point: 106 °C
  • Solubility: Soluble in water, slightly soluble in ethanol, insoluble in ether
  • Storage: Room temperature (recommended <15 °C); store under inert gas; hygroscopic

Catalog No. 50-278-1437 Supplier Grainger Supplier No. 19KR25
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-(+)-Maltose Monohydrate is a disaccharide sugar composed of two glucose units linked by an α(1→4) glycosidic bond. This white, crystalline powder is suitable for use in research, brewing, and fermentation processes as a carbohydrate source.

  • Formula: C12H22O11·H2O
  • Molecular weight: 360.32
  • Purity: ≥98.0% (HPLC)
  • Physical state: White powder to crystal
  • Melting point: 106 °C
  • Solubility: Soluble in water, slightly soluble in ethanol, insoluble in ether
  • Storage: Room temperature (recommended <15 °C); store under inert gas; hygroscopic

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.