Learn More
Alkali Scientific D (+) Maltose, Monohydrate, 500 G
SDP
Catalog No. 5000660429
Encompass_Preferred
D (+) Maltose, Monohydrate, 500 G is a pure carbohydrate reagent used in biochemical, pharmaceutical, and food testing applications. Maltose is a disaccharide, often used as a substrate in fermentation processes and as an energy source in microbial and enzymatic assays. The 500G size ensures sufficient quantities for larger-scale experiments, making it suitable for research labs and production environments. This reagent is commonly used in glucose metabolism studies, microbial fermentation, and enzyme activity testing. The monohydrate form maintains stability, ensuring accurate, reproducible results. Maltose also plays an essential role in food science, particularly in the testing of sugar-related products and the development of new formulations.
Catalog No. 50-006-60429 Supplier Alkali Scientific Supplier No. ALT003CP
D (+) Maltose, Monohydrate, 500 G is a pure carbohydrate reagent used in biochemical, pharmaceutical, and food testing applications. Maltose is a disaccharide, often used as a substrate in fermentation processes and as an energy source in microbial and enzymatic assays. The 500G size ensures sufficient quantities for larger-scale experiments, making it suitable for research labs and production environments. This reagent is commonly used in glucose metabolism studies, microbial fermentation, and enzyme activity testing. The monohydrate form maintains stability, ensuring accurate, reproducible results. Maltose also plays an essential role in food science, particularly in the testing of sugar-related products and the development of new formulations.