Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals D-(+)-Maltose monohydrate, 95%

Catalog No. AAA1626636
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1626636 500 g
AAA1626622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1626636 Supplier Thermo Scientific Chemicals Supplier No. A1626636
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 6363-53-7 | C12H24O12 | 360.31 g/mol

D-(+)-Maltose monohydrate, is commonly found in foods and commonly utilized in brewing processes. It is also used in various culture media in the cell and tissue culture applications. D-(+)-Maltose Monohydrate is used as a substrate for α-glucosidase. It is also used as a substrate for the identification, differentiation and characterization of enzymes such as maltase(s); maltose α-D-glucosyltransferase(s); maltose-transporting ATPase(s); maltose O-acetyltransferase(s) and maltose epimerase(s) and phosphorylase(s). D-Maltose is used to study maltose-binding proteins and disaccharide transport systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(+)-Maltose monohydrate, is commonly found in foods and commonly utilized in brewing processes. It is also used in various culture media in the cell and tissue culture applications. D-(+)-Maltose Monohydrate is used as a substrate for α-glucosidase. It is also used as a substrate for the identification, differentiation and characterization of enzymes such as maltase(s); maltose α-D-glucosyltransferase(s); maltose-transporting ATPase(s); maltose O-acetyltransferase(s) and maltose epimerase(s) and phosphorylase(s). D-Maltose is used to study maltose-binding proteins and disaccharide transport systems.

Solubility
Soluble in water. Slightly soluble in alcohol. Insoluble in ether

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6363-53-7
Molecular Formula C12H24O12
Molecular Weight (g/mol) 360.31
MDL Number MFCD00149343
InChI Key HBDJFVFTHLOSDW-UHFFFAOYNA-N
Synonym d-+-maltose monohydrate, unii-dm477ee40d, 2r,3r,4r,5r-2,3,5,6-tetrahydroxy-4-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy hexanal hydrate, beta-maltose monohydrate, d-+-maltosemonohydrate, 69-79-4 anhydrous, d +-maltose monohydrate, d-+-maltose monohydrate, puriss., d-+-maltose monohydrate, analytical standard, d-+-maltose monohydrate, bioxtra
IUPAC Name 2,3,5,6-tetrahydroxy-4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}hexanal hydrate
SMILES O.OCC(O)C(OC1OC(CO)C(O)C(O)C1O)C(O)C(O)C=O

Specifications

Melting Point ∼120°C (decomposition)
Assay Percent Range 95%
Quantity 500 g
Beilstein 5784659
Merck Index 14,5714
Solubility Information Soluble in water; Slightly soluble in alcohol; Insoluble in ether
Optical Rotation +130° (c=2 in Water, 24h)
Formula Weight 360.32 (342.30 Anhydrous)
Percent Purity 95%
Chemical Name or Material D-(+)-Maltose monohydrate
EINECSNumber 200-716-5
RTECSNumber OO5250000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.