Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-(+)-Maltose Monohydrate, ∼99%, MP Biomedicals™

Catalog No. 0219023791 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-190-23791 1 kg
ICN19023790 500 g
ICN19023780 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02-190-23791 Supplier MP Biomedicals, Inc Supplier No. 0219023791
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-(+)-Maltose Monohydrate, ∼99% is a sweetener.

  • D-(+)-Maltose monohydrate is used as a sweetener with about one-third the sweetness of sucrose and as a nutrient in culture media.
  • It is used in pharmaceutical formulations and as a parenteral supplement of sugar for diabetics.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6363-53-7
Molecular Formula C12H24O12
Molecular Weight (g/mol) 360.31
MDL Number MFCD00149343
InChI Key HBDJFVFTHLOSDW-UHFFFAOYNA-N
Synonym d-+-maltose monohydrate, unii-dm477ee40d, 2r,3r,4r,5r-2,3,5,6-tetrahydroxy-4-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy hexanal hydrate, beta-maltose monohydrate, d-+-maltosemonohydrate, 69-79-4 anhydrous, d +-maltose monohydrate, d-+-maltose monohydrate, puriss., d-+-maltose monohydrate, analytical standard, d-+-maltose monohydrate, bioxtra
IUPAC Name 2,3,5,6-tetrahydroxy-4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}hexanal hydrate
SMILES O.OCC(O)C(OC1OC(CO)C(O)C(O)C1O)C(O)C(O)C=O

Specifications

Color White
Density 1.54 g/mL
Assay Percent Range 99%
Quantity 1 kg
Optical Rotation 130° ± 5°(c=4, H2O, NH3OH)
Formula Weight 360.31
Percent Purity 99%
Physical Form Powder
Chemical Name or Material D-(+)-Maltose monohydrate, Molecular biology
Recommended Storage Store at room temperature.

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.