Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-(+)-Maltose monohydrate, MilliporeSigma™ Supelco™

Catalog No. 111015000 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5000 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5000 Supplier MilliporeSigma Supelco Supplier No. 47288
Add to Cart
Add to Cart
Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

CAS 6363-53-7
Physical Form Neat
Linear Formula C12H22O11 · H2O
MDL Number MFCD00149343
Synonym 4-O-alpha-D-Glucopyranosyl-D-glucose; Maltobiose
Type Analytical Standard
Quantity 500 mg
Formula Weight 360.31
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.