Learn More
American Radiolabeled Chemicals Inc D-MAN 14CU 50UC
SDP
Catalog No. 50353058
Encompass
Mannose, D-14C(U) 50uCi in ethanol:water (9:1), 0.1 mCi/ml, 250-360 mCi/mmol, M.W. 180.2, Exclusive
Catalog No. 50-353-058 Supplier American Radiolabeled Chemicals Inc Supplier No. ARC 357550UC
Mannose, D-14C(U) 50uCi in ethanol:water (9:1), 0.1 mCi/ml, 250-360 mCi/mmol, M.W. 180.2, Exclusive