Learn More
Medchemexpress LLC D-(-)-mandelic acid | 611-71-2 | MFCD00064251 | 99.9% | 152.15 g/mol | C8H8O3 | 100g
SDP
Catalog No. 5000269284
Encompass_Preferred
D-(-)-Mandelic acid is an orally active alpha hydroxycarboxylic acid that can be isolated from bitter almonds and Indian chestnut trees It has antioxidant and antibacterial properties and is expected to play an important role in the treatment of rheumatoid arthritis[1][4] /br
Catalog No. 50-002-69284 Supplier Medchemexpress LLC Supplier No. HYY0585100G
D-(-)-Mandelic acid is an orally active alpha hydroxycarboxylic acid that can be isolated from bitter almonds and Indian chestnut trees It has antioxidant and antibacterial properties and is expected to play an important role in the treatment of rheumatoid arthritis[1][4] /br