Learn More
Medchemexpress LLC D-mannitol-13C6 | 287112-34-9 | MFCD01075541 | 98.0% | 188.13 g/mol | 13C6H14O6 | 10 MG
SDP
Catalog No. 5000296370
Encompass_Preferred
D-mannitol-13C6 is the uniformly 13C-labeled form of D-mannitol supplied as a solid stable-isotope standard for analytical and biochemical applications. It is used as an internal or tracer standard for mass spectrometry and metabolic tracing, and functions as an osmotic agent in biological experiments.
  • Uniform 13C labeling for accurate isotope tracing and quantitation.
  • High purity (98.0%) suitable for analytical applications.
  • Supplied as a stable solid for convenient handling and storage.
  • Available in small, precise quantities for trace-level experiments.
  • Recommended storage conditions support long-term stability.
Catalog No. 50-002-96370 Supplier Medchemexpress LLC Supplier No. HYN0378S210MG
D-mannitol-13C6 is the uniformly 13C-labeled form of D-mannitol supplied as a solid stable-isotope standard for analytical and biochemical applications. It is used as an internal or tracer standard for mass spectrometry and metabolic tracing, and functions as an osmotic agent in biological experiments.
  • Uniform 13C labeling for accurate isotope tracing and quantitation.
  • High purity (98.0%) suitable for analytical applications.
  • Supplied as a stable solid for convenient handling and storage.
  • Available in small, precise quantities for trace-level experiments.
  • Recommended storage conditions support long-term stability.