Learn More
Apexbio Technology LLC D-Mannitol 69-65-8 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062153
Encompass_Preferred
D-Mannitol is a six-carbon sugar alcohol commonly used in biomedical research as an osmotic regulator Due to its cell membrane impermeability this molecule creates an extracellular osmotic gradient facilitating transmembrane water movement and influencing fluid distribution between intracellular and extracellular compartments D-Mannitol s mode of action involves modulating osmotic pressure enabling applications in cell cryopreservation blood-brain barrier permeability investigations intracranial pressure regulation and drug delivery system development It is suitable for cellular cultivation physiological and pharmacological experiments as well as foundational and applied biopharmaceutical research
Catalog No. 50-000-62153 Supplier Apexbio Technology LLC Supplier No. B20905.1
D-Mannitol is a six-carbon sugar alcohol commonly used in biomedical research as an osmotic regulator Due to its cell membrane impermeability this molecule creates an extracellular osmotic gradient facilitating transmembrane water movement and influencing fluid distribution between intracellular and extracellular compartments D-Mannitol s mode of action involves modulating osmotic pressure enabling applications in cell cryopreservation blood-brain barrier permeability investigations intracranial pressure regulation and drug delivery system development It is suitable for cellular cultivation physiological and pharmacological experiments as well as foundational and applied biopharmaceutical research