Learn More
Sigma Aldrich Fine Chemicals Biosciences D-Mannitol, 69-65-8, MFCD00064287, 500 G
Catalog No. NC0969332
Encompass
Empirical Formula (Hill Notation): C6H14O6, Molecular Weight: 182.17, ≥98%, Synonym: Mannite. D-mannitol has been used as a hydroxyl radical scavenger and as a medium supplement.
Catalog No. NC0969332 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. M4125500G
Empirical Formula (Hill Notation): C6H14O6, Molecular Weight: 182.17, ≥98%, Synonym: Mannite. D-mannitol has been used as a hydroxyl radical scavenger and as a medium supplement.