Learn More
Medchemexpress LLC D-Mannitol | 69-65-8 | 182.17 | 1 ML
SDP
Catalog No. 5000389315
Encompass_Preferred
D-Mannitol is an oral, resistant sugar widely used in the food and pharmaceutical industries. It acts as an osmotic diuretic, promoting the absorption and retention of calcium and magnesium, and reducing tissue edema. This compound can also enhance brown fat formation, improve insulin effects, and reduce blood sugar levels.
  • Promotes absorption and retention of calcium and magnesium
  • Reduces tissue edema as an osmotic diuretic
  • Enhances brown fat formation and improves insulin effect
  • Reduces blood sugar levels
  • Induces white fat cells to become brown fat cells
  • Maintains osmotic pressure between plant cytoplasm and culture medium
  • Protects cells when the cell wall is weakened or removed
  • Functions as a weak renal vasodilator
Catalog No. 50-003-89315 Supplier Medchemexpress LLC Supplier No. HYN037810MM/1ML
D-Mannitol is an oral, resistant sugar widely used in the food and pharmaceutical industries. It acts as an osmotic diuretic, promoting the absorption and retention of calcium and magnesium, and reducing tissue edema. This compound can also enhance brown fat formation, improve insulin effects, and reduce blood sugar levels.
  • Promotes absorption and retention of calcium and magnesium
  • Reduces tissue edema as an osmotic diuretic
  • Enhances brown fat formation and improves insulin effect
  • Reduces blood sugar levels
  • Induces white fat cells to become brown fat cells
  • Maintains osmotic pressure between plant cytoplasm and culture medium
  • Protects cells when the cell wall is weakened or removed
  • Functions as a weak renal vasodilator