Learn More
AdipoGen D-Mannitol
SDP
Catalog No. 502560468
Encompass_Preferred
Chemical. CAS 69-65-8. Formula C6H14O6. MW 182.17. Synthetic. Sugar alcohol that can be used as an inert osmotic control substance. The uptake and phosphorylation of D-Mannitol is catalyzed by the mannitol-specific phosphoenolpyruvate-dependent phosphotransferase systems PTS. Used in pharmacy as excipient and diluent for solids and liquids and in analytical chemistry for boron determinations. It is used as an anticaking and free-flow agent, flavoring agent, lubricant and release agent, stabilizer and thickener and nutritive sweetener in the food industry. Used in sweetness inhibition studies. Can interact with neutrophils and monocytes. Experiments have shown that it is able to decrease neutrophil apoptosis in vitro. Can lower fat digestibility and body fat accumulation in rats, as well as upregulate monocyte HLA-DR, monocyte and neutrophil CD11b.
Catalog No. 50-256-0468 Supplier AdipoGen Supplier No. CDXM0228G500
Chemical. CAS 69-65-8. Formula C6H14O6. MW 182.17. Synthetic. Sugar alcohol that can be used as an inert osmotic control substance. The uptake and phosphorylation of D-Mannitol is catalyzed by the mannitol-specific phosphoenolpyruvate-dependent phosphotransferase systems PTS. Used in pharmacy as excipient and diluent for solids and liquids and in analytical chemistry for boron determinations. It is used as an anticaking and free-flow agent, flavoring agent, lubricant and release agent, stabilizer and thickener and nutritive sweetener in the food industry. Used in sweetness inhibition studies. Can interact with neutrophils and monocytes. Experiments have shown that it is able to decrease neutrophil apoptosis in vitro. Can lower fat digestibility and body fat accumulation in rats, as well as upregulate monocyte HLA-DR, monocyte and neutrophil CD11b.