Learn More
Medchemexpress LLC D-Mannitol (Standard) | 69-65-8 | MFCD00064287 | 182.17 | 50 MG
SDP
Catalog No. 5000390089
Encompass_Preferred
D-Mannitol (Standard) is the analytical standard of D-Mannitol, intended for research and analytical applications. It is an oral, resistant sugar widely used in the food and pharmaceutical industries. It promotes the absorption and retention of calcium and magnesium through cecal fermentation and acts as an osmotic diuretic to reduce tissue edema.
  • Enhances brown fat formation.
  • Improves insulin effect and reduces blood sugar levels.
  • Induces white fat cells into brown fat cells by starting the β3-adrenergic receptor (β3-AR), PGC1α, and PKA.
  • Maintains osmotic pressure between the plant cytoplasm and the culture medium.
  • Protects cells when the cell wall is weakened or removed.
  • Used in qualitative, quantitative, and methodological research experiments in techniques like HPLC, GC, and MS.
Catalog No. 50-003-90089 Supplier Medchemexpress LLC Supplier No. HYN0378R50MG
D-Mannitol (Standard) is the analytical standard of D-Mannitol, intended for research and analytical applications. It is an oral, resistant sugar widely used in the food and pharmaceutical industries. It promotes the absorption and retention of calcium and magnesium through cecal fermentation and acts as an osmotic diuretic to reduce tissue edema.
  • Enhances brown fat formation.
  • Improves insulin effect and reduces blood sugar levels.
  • Induces white fat cells into brown fat cells by starting the β3-adrenergic receptor (β3-AR), PGC1α, and PKA.
  • Maintains osmotic pressure between the plant cytoplasm and the culture medium.
  • Protects cells when the cell wall is weakened or removed.
  • Used in qualitative, quantitative, and methodological research experiments in techniques like HPLC, GC, and MS.