Learn More
Medchemexpress LLC D-Mannoheptulose | 3615-44-9 | MFCD00006622 | 210.18 | 10 MG
SDP
Catalog No. 5000447566
Encompass_Preferred
D-Mannoheptulose is a 7-carbon keto sugar found in avocado. It is a specific inhibitor of D-glucose phosphorylation by hexokinase isoenzymes. This compound can block insulin release and the utilization of carbohydrates in rats. It also exhibits various functions such as antioxidant and anticancer effects, and can act as an energy source.
  • Specific inhibitor of D-glucose phosphorylation by hexokinase isoenzymes
  • Can block insulin release
  • Can block utilization of carbohydrates in rats
  • Exhibits antioxidant effects
  • Shows anticancer effects
  • Acts as an energy source
  • Inhibits breast cancer cell line growth (MCF-7 IC50: 263.3 μg/mL)
  • No significant cytotoxic effect on normal cells (HMECs IC50: 975.1 μg/mL)
Catalog No. 50-004-47566 Supplier Medchemexpress LLC Supplier No. HYU0046210MG
D-Mannoheptulose is a 7-carbon keto sugar found in avocado. It is a specific inhibitor of D-glucose phosphorylation by hexokinase isoenzymes. This compound can block insulin release and the utilization of carbohydrates in rats. It also exhibits various functions such as antioxidant and anticancer effects, and can act as an energy source.
  • Specific inhibitor of D-glucose phosphorylation by hexokinase isoenzymes
  • Can block insulin release
  • Can block utilization of carbohydrates in rats
  • Exhibits antioxidant effects
  • Shows anticancer effects
  • Acts as an energy source
  • Inhibits breast cancer cell line growth (MCF-7 IC50: 263.3 μg/mL)
  • No significant cytotoxic effect on normal cells (HMECs IC50: 975.1 μg/mL)