Learn More
AdipoGen D-Mannoheptulose
SDP
Catalog No. 502562210
Encompass_Preferred
Chemical. CAS 3615-44-9. Formula C7H14O7. MW 210.2. . Naturally occurring heptose in avocado fruit. Hexokinase HK and glucokinase inhibitor in diverse organisms by competing with D-glucose. Useful agent for immunometabolism research. Blocks glucose phosphorylation by blocking hexokinase. Prevents the conversion of glucose to glucose-6-phosphate that can mediate the activation of the carbohydrate response element binding protein and consequently inhibiting the rate limiting step of glycolysis. Blocks glucose oxidation and glucose-mediated insulin release in pancreatic islet cells.
Catalog No. 50-256-2210 Supplier AdipoGen Supplier No. AGCR13695M005
Chemical. CAS 3615-44-9. Formula C7H14O7. MW 210.2. . Naturally occurring heptose in avocado fruit. Hexokinase HK and glucokinase inhibitor in diverse organisms by competing with D-glucose. Useful agent for immunometabolism research. Blocks glucose phosphorylation by blocking hexokinase. Prevents the conversion of glucose to glucose-6-phosphate that can mediate the activation of the carbohydrate response element binding protein and consequently inhibiting the rate limiting step of glycolysis. Blocks glucose oxidation and glucose-mediated insulin release in pancreatic islet cells.