Learn More
Medchemexpress LLC D-mannopyranose, 1,2,3,4,6-penta-O-acetyl | 25941-03-1 | MFCD05864874 | 98.0% | 390.34 | C16H22O11 | 25 G
SDP
Catalog No. 5000303714
Encompass_Preferred
1,2,3,4,6-Penta-O-acetyl-D-mannopyranose is an acetyl-protected derivative of D-mannopyranose used as a synthetic intermediate in carbohydrate chemistry. It is supplied as a solid with molecular formula C16H22O11 and molecular weight 390.34 g/mol. Typical applications include glycoside synthesis and preparative carbohydrate modifications.
  • Acetyl-protected hydroxyl groups facilitate selective glycosylation reactions.
  • Suitable for synthesis of tetra-O-acetyl-O-prop-1-yl-α-D-mannopyranoside and related derivatives.
  • Available in small pack sizes appropriate for laboratory synthesis.
  • Characterized by standard chemical identifiers and documented analytical data.
  • Supplied with batch-specific certificate of analysis for quality verification.
Catalog No. 50-003-03714 Supplier Medchemexpress LLC Supplier No. 25GHYW039899
1,2,3,4,6-Penta-O-acetyl-D-mannopyranose is an acetyl-protected derivative of D-mannopyranose used as a synthetic intermediate in carbohydrate chemistry. It is supplied as a solid with molecular formula C16H22O11 and molecular weight 390.34 g/mol. Typical applications include glycoside synthesis and preparative carbohydrate modifications.
  • Acetyl-protected hydroxyl groups facilitate selective glycosylation reactions.
  • Suitable for synthesis of tetra-O-acetyl-O-prop-1-yl-α-D-mannopyranoside and related derivatives.
  • Available in small pack sizes appropriate for laboratory synthesis.
  • Characterized by standard chemical identifiers and documented analytical data.
  • Supplied with batch-specific certificate of analysis for quality verification.