Learn More
Medchemexpress LLC D-Mannose-13C | 70849-31-9 | 181.15 | 50 MG
SDP
Catalog No. 5000389839
Encompass_Preferred
D-Mannose-13C is the 13C labeled D-Mannose. D-Mannose is a carbohydrate that plays an important role in human metabolism, especially in the glycosylation of specific proteins.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • For research use only.
Catalog No. 50-003-89839 Supplier Medchemexpress LLC Supplier No. HYN0379S550MG
D-Mannose-13C is the 13C labeled D-Mannose. D-Mannose is a carbohydrate that plays an important role in human metabolism, especially in the glycosylation of specific proteins.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • For research use only.