Learn More
Medchemexpress LLC D-Mannose-13C6 | 287100-74-7 | 186.11 | 1 MG
SDP
Catalog No. 5000389147
Encompass_Preferred
D-Mannose-13C6 is a carbohydrate that plays an important role in human metabolism, especially in the glycosylation of specific proteins. It is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-89147 Supplier Medchemexpress LLC Supplier No. HYN0379S221MG
D-Mannose-13C6 is a carbohydrate that plays an important role in human metabolism, especially in the glycosylation of specific proteins. It is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.