Learn More
Medchemexpress LLC D-Mannose-13C6 | 287100-74-7 | 186.11 | 5 MG
SDP
Catalog No. 5000389339
Encompass_Preferred
D-Mannose-13C6 is a carbohydrate that plays a crucial role in human metabolism, particularly in the glycosylation of specific proteins. It is intended for research use only.
  • This compound can be used as a tracer.
  • It can serve as an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes, such as carbon-13, are incorporated into drug molecules, primarily as tracers for quantitation during drug development.
  • Deuteration, a process involving stable heavy isotopes, has gained attention for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-89339 Supplier Medchemexpress LLC Supplier No. HYN0379S225MG
D-Mannose-13C6 is a carbohydrate that plays a crucial role in human metabolism, particularly in the glycosylation of specific proteins. It is intended for research use only.
  • This compound can be used as a tracer.
  • It can serve as an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes, such as carbon-13, are incorporated into drug molecules, primarily as tracers for quantitation during drug development.
  • Deuteration, a process involving stable heavy isotopes, has gained attention for its potential to influence the pharmacokinetic and metabolic profiles of drugs.