Learn More
TARGETMOL CHEMICALS INC D-Mannose 1G
SDP
Catalog No. 502879978
Encompass_Preferred
Also available in 1 mL, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. D-Mannose (D-(+)-Mannose) is a carbohydrate. D-Mannose plays an important role in human metabolism, especially in the glycosylation of specific proteins. Purity 99.21%
Catalog No. 50-287-9978 Supplier TARGETMOL CHEMICALS INC Supplier No. T48121G
Also available in 1 mL, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. D-Mannose (D-(+)-Mannose) is a carbohydrate. D-Mannose plays an important role in human metabolism, especially in the glycosylation of specific proteins. Purity 99.21%