Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-Mannose | 3458-28-4 | 180.16 | 25 MG
SDP

Catalog No. 5000389633
Encompass_Preferred

D-Mannose (Standard) is the analytical standard of D-Mannose, intended for research and analytical applications. It is a C-2 superpolymer of glucose found naturally in many plants and fruits. D-Mannose exhibits anti-inflammatory and antitumor activity and plays a significant role in immune regulation.

  • Purity: 99.77%
  • Application: the compound is the grade of analytical standard, which is the reference standard supplied assay. It is commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
  • Appearance: solid
  • Color: white to off-white
  • Structure classification: saccharides
  • Initial source: endogenous metabolite, human gut microbiota metabolites
  • Shipping: room temperature in continental US; may vary elsewhere.
  • Storage: please store the product under the recommended conditions in the Certificate of Analysis.

Catalog No. 50-003-89633 Supplier Medchemexpress LLC Supplier No. 25MGHYN0379R
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Mannose (Standard) is the analytical standard of D-Mannose, intended for research and analytical applications. It is a C-2 superpolymer of glucose found naturally in many plants and fruits. D-Mannose exhibits anti-inflammatory and antitumor activity and plays a significant role in immune regulation.

  • Purity: 99.77%
  • Application: the compound is the grade of analytical standard, which is the reference standard supplied assay. It is commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
  • Appearance: solid
  • Color: white to off-white
  • Structure classification: saccharides
  • Initial source: endogenous metabolite, human gut microbiota metabolites
  • Shipping: room temperature in continental US; may vary elsewhere.
  • Storage: please store the product under the recommended conditions in the Certificate of Analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.