Learn More
Medchemexpress LLC D-Mannose | 3458-28-4 | MFCD00064122 | 180.16 | 1 G
SDP
Catalog No. 5000390060
Encompass_Preferred
D-Mannose is a C-2 superpolymer of glucose that occurs naturally in many plants and fruits. It is intended for research use only.
  • Exhibits anti-inflammatory and antitumor activity
  • Plays an important role in immune regulation
  • Inhibits macrophage activation by inhibiting IL-1β production
  • Stimulates Treg cell differentiation by promoting TGF-β activation
  • Protects against DSS-induced ulcerative colitis in mice
  • Alleviates bone loss in mice through Treg cell proliferation and intestinal microbiome-dependent anti-inflammatory effects
Catalog No. 50-003-90060 Supplier Medchemexpress LLC Supplier No. HYN03791G
D-Mannose is a C-2 superpolymer of glucose that occurs naturally in many plants and fruits. It is intended for research use only.
  • Exhibits anti-inflammatory and antitumor activity
  • Plays an important role in immune regulation
  • Inhibits macrophage activation by inhibiting IL-1β production
  • Stimulates Treg cell differentiation by promoting TGF-β activation
  • Protects against DSS-induced ulcerative colitis in mice
  • Alleviates bone loss in mice through Treg cell proliferation and intestinal microbiome-dependent anti-inflammatory effects