Learn More
Medchemexpress LLC D-Mannose | 3458-28-4 | MFCD00064122 | 180.16 | 1 ML
SDP
Catalog No. 5000389338
Encompass_Preferred
D-Mannose is a C-2 superpolymer of glucose, naturally occurring in various plants and fruits, exhibiting both anti-inflammatory and antitumor properties. It is an endogenous metabolite and a carbohydrate that plays a crucial role in human metabolism.
  • Endogenous metabolite
  • Carbohydrate involved in human metabolism
  • Important for glycosylation of specific proteins
  • Exhibits anti-inflammatory activity
  • Shows antitumor activity
  • Supplied as a 10 mM solution in DMSO
Catalog No. 50-003-89338 Supplier Medchemexpress LLC Supplier No. HYN037910MM/1ML
D-Mannose is a C-2 superpolymer of glucose, naturally occurring in various plants and fruits, exhibiting both anti-inflammatory and antitumor properties. It is an endogenous metabolite and a carbohydrate that plays a crucial role in human metabolism.
  • Endogenous metabolite
  • Carbohydrate involved in human metabolism
  • Important for glycosylation of specific proteins
  • Exhibits anti-inflammatory activity
  • Shows antitumor activity
  • Supplied as a 10 mM solution in DMSO