Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-mannose | 3458-28-4 | MFCD00799233 | 99.8% | 180.16 g/mol | C6H12O6 | 10 MG
SDP

Catalog No. 5000295836
Encompass_Preferred

D-Mannose is an analytical reference standard supplied as a white to off-white solid monosaccharide for use in qualitative and quantitative research, method development, and verification by techniques such as HPLC, GC, and mass spectrometry.

  • Intended for research and analytical applications.
  • Purity: 99.8%.
  • Chemical formula: C6H12O6.
  • Molecular weight: 180.16 g/mol.
  • CAS number: 3458-28-4.
  • Appearance: white to off-white solid.
  • Includes a 10 MG pack; other pack sizes are available.

Catalog No. 50-002-95836 Supplier Medchemexpress LLC Supplier No. HYN0379R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Mannose is an analytical reference standard supplied as a white to off-white solid monosaccharide for use in qualitative and quantitative research, method development, and verification by techniques such as HPLC, GC, and mass spectrometry.

  • Intended for research and analytical applications.
  • Purity: 99.8%.
  • Chemical formula: C6H12O6.
  • Molecular weight: 180.16 g/mol.
  • CAS number: 3458-28-4.
  • Appearance: white to off-white solid.
  • Includes a 10 MG pack; other pack sizes are available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.