Learn More
Alkali Scientific D (+) Mannose, 500 Grams
SDP
Catalog No. 5000660623
Encompass_Preferred
D (+) Mannose, 500 G, is a pure form of the sugar mannose used as a reagent in various biochemical and medical research applications. Mannose is important for studying carbohydrate metabolism, particularly in the context of glycosylation and the effects of mannose on cellular signaling. This compound is frequently used in cell culture experiments, particularly in the preparation of media for organisms that metabolize mannose. It is also utilized in clinical research studies focused on the role of mannose in immune system modulation and the treatment of diseases such as mannose-binding lectin deficiency. The 500g size is suitable for large-scale research applications.
Catalog No. 50-006-60623 Supplier Alkali Scientific Supplier No. ANN001CP
D (+) Mannose, 500 G, is a pure form of the sugar mannose used as a reagent in various biochemical and medical research applications. Mannose is important for studying carbohydrate metabolism, particularly in the context of glycosylation and the effects of mannose on cellular signaling. This compound is frequently used in cell culture experiments, particularly in the preparation of media for organisms that metabolize mannose. It is also utilized in clinical research studies focused on the role of mannose in immune system modulation and the treatment of diseases such as mannose-binding lectin deficiency. The 500g size is suitable for large-scale research applications.