Learn More
AdipoGen D Mannose
SDP
Supplier:  AdipoGen CDXM0020G025
Encompass_Preferred
Chemical. CAS 3458-28-4. Formula C6H12O6. MW 180.16. Synthetic. Mannose is an aldohexose carbohydrate. This product may be used to study the effects of exogenous mannose on in vitro processes such as glycosylation.
Catalog No. 50-256-0563