Learn More
Medchemexpress LLC α-D-mannose pentaacetate | 4163-65-9 | MFCD00069798 | 97.0% | 390.34 | C16H22O11 | 100 G
SDP
Catalog No. 5000299828
Encompass_Preferred
α-D-mannose pentaacetate is the pentaacetylated ester of α-D-mannose used as a biochemical reagent and intermediate in carbohydrate chemistry. It is supplied as a solid reagent for glycosylation, synthetic transformations, and analytical applications.
  • Acetylated monosaccharide for glycosylation and carbohydrate synthesis.
  • High purity suitable for research and analytical use.
  • Solid formulation simplifies storage and weighing.
  • Useful as a synthetic intermediate and reference standard.
  • Molecular formula C16H22O11 and molecular weight 390.34 support experimental planning.
Catalog No. 50-002-99828 Supplier Medchemexpress LLC Supplier No. HYW039898100G
α-D-mannose pentaacetate is the pentaacetylated ester of α-D-mannose used as a biochemical reagent and intermediate in carbohydrate chemistry. It is supplied as a solid reagent for glycosylation, synthetic transformations, and analytical applications.
  • Acetylated monosaccharide for glycosylation and carbohydrate synthesis.
  • High purity suitable for research and analytical use.
  • Solid formulation simplifies storage and weighing.
  • Useful as a synthetic intermediate and reference standard.
  • Molecular formula C16H22O11 and molecular weight 390.34 support experimental planning.