Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC α-D-mannose pentaacetate | 4163-65-9 | MFCD00069798 | 97.0% | 390.34 g/mol | C16H22O11 | 25 G
SDP

Catalog No. 5000303416
Encompass_Preferred

α-D-Mannose pentaacetate is an acetylated derivative of D-mannose used as a biochemical reagent and building block in carbohydrate chemistry. It is suitable for glycosylation studies and other life-science research that require protected sugar derivatives.

  • Acetylated D-mannose derivative for glycosylation and synthesis studies.
  • High purity: 97.0%.
  • Physical form: solid, white to off-white.
  • Molecular formula C16H22O11; molecular weight 390.34 g/mol.
  • Storage: powder at -20°C (3 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Available pack sizes: 25 g, 50 g, 100 g, bulk on inquiry.

Catalog No. 50-003-03416 Supplier Medchemexpress LLC Supplier No. 25GHYW039898
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

α-D-Mannose pentaacetate is an acetylated derivative of D-mannose used as a biochemical reagent and building block in carbohydrate chemistry. It is suitable for glycosylation studies and other life-science research that require protected sugar derivatives.

  • Acetylated D-mannose derivative for glycosylation and synthesis studies.
  • High purity: 97.0%.
  • Physical form: solid, white to off-white.
  • Molecular formula C16H22O11; molecular weight 390.34 g/mol.
  • Storage: powder at -20°C (3 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Available pack sizes: 25 g, 50 g, 100 g, bulk on inquiry.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.