Learn More
Medchemexpress LLC D-Mannose (Standard) | 3458-28-4 | 180.16 | 50 MG
SDP
Catalog No. 5000390088
Encompass_Preferred
D-Mannose (Standard) is an analytical standard used in research and analytical applications. This C-2 superpolymer of glucose occurs naturally in many plants and fruits, exhibiting anti-inflammatory and antitumor activity and playing an important role in immune regulation. It serves as a reference standard in qualitative, quantitative, and methodological research experiments.
  • Suitable for HPLC, GC, and MS applications
  • Purity of 99.77%
  • Appears as a solid, white to off-white substance
  • Derived from endogenous metabolite, human gut microbiota metabolites, and microorganisms
Catalog No. 50-003-90088 Supplier Medchemexpress LLC Supplier No. HYN0379R50MG
D-Mannose (Standard) is an analytical standard used in research and analytical applications. This C-2 superpolymer of glucose occurs naturally in many plants and fruits, exhibiting anti-inflammatory and antitumor activity and playing an important role in immune regulation. It serves as a reference standard in qualitative, quantitative, and methodological research experiments.
  • Suitable for HPLC, GC, and MS applications
  • Purity of 99.77%
  • Appears as a solid, white to off-white substance
  • Derived from endogenous metabolite, human gut microbiota metabolites, and microorganisms