Learn More
Medchemexpress LLC HY-N2340 100mg Medchemexpress, D-(+)-Melezitose CAS:597-12-6 Purity:>98%
SDP
Catalog No. 501933919
Encompass_Preferred
Medchemexpress, HY-N2340 100mg D-(+)-Melezitose CAS:597-12-6 D-(+)-Melezitose can be used to identify clinical isolates of indole-positive and indole-negative Klebsiella spp. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3919 Supplier Medchemexpress LLC Supplier No. HYN2340100MG
Medchemexpress, HY-N2340 100mg D-(+)-Melezitose CAS:597-12-6 D-(+)-Melezitose can be used to identify clinical isolates of indole-positive and indole-negative Klebsiella spp. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.